Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 8, 9 | 1225 | ||
NM_001271900.1 | NP_001258829.1 | 9, 10 | 1419 | |||
NM_001271948.1 | NP_001258877.1 | 8, 9 | 1752 | |||
NM_181674.2 | NP_858060.2 | 8, 9 | 1550 | |||
NM_181675.3 | NP_858061.2 | 8, 9 | 1272 | |||
NM_181676.2 | NP_858062.1 | 7, 8 | 1356 | |||
NM_181677.2 | NP_858063.1 | 9, 10 | 1182 | |||
NM_181678.2 | NP_858064.1 | 9, 10 | 1195 | |||
NR_073526.1 | 7, 8 | 1667 | ||||
NR_073527.1 | 8, 9 | 1304 | ||||
GenBank mRNA | AK056192.1 | - | 8, 9 | 1232 | ||
AK289717.1 | - | 8, 9 | 1201 | |||
AK294659.1 | - | 9, 10 | 1328 | |||
AK295347.1 | - | 8, 9 | 1196 | |||
AK314810.1 | - | 8, 9 | 1549 | |||
BC031790.1 | - | 8, 9 | 1440 | 4939981 | ||
BX647887.1 | - | 8, 9 | 1286 | |||
M64930.1 | - | 7, 8 | 1478 |