Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr16c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080710.2 | NP_001074179.1 | 2 | 393 | ||
GenBank mRNA | AK076465.1 | - | 2 | 374 | ||
BC125450.1 | - | 2 | 393 | 40129965 | ||
BC125452.1 | - | 2 | 393 | 40129967 |