Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172868.3 | NP_766456.1 | 6 | 1010 | ||
GenBank mRNA | AK045344.1 | - | 6 | 1011 | ||
BC113156.1 | - | 6 | 976 | 40058559 |
Gene Symbol: Pakap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF064781.1 | - | 1 | 132 |