Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 3, 4 | 553 | ||
NM_002718.4 | NP_002709.2 | 4, 5 | 2988 | |||
NM_181897.2 | NP_871626.1 | 3, 4 | 843 | |||
XM_006713686.3 | XP_006713749.1 | 4, 5 | 3399 | |||
XM_011512956.2 | XP_011511258.1 | 4, 5 | 3163 | |||
XM_017006787.1 | XP_016862276.1 | 3, 4 | 2721 | |||
GenBank mRNA | AK293012.1 | - | 4, 5 | 2968 | ||
AK298059.1 | - | 3, 4 | 482 | |||
AK299544.1 | - | 3, 4 | 513 | |||
AK302163.1 | - | 2, 3 | 251 | |||
AK314243.1 | - | 4, 5 | 2916 | |||
AK316258.1 | - | 3, 4 | 796 | |||
AL389975.1 | - | 1, 2 | 71 | |||
BC065531.1 | - | 4, 5 | 2935 | 6161260 | ||
L07590.1 | - | 4, 5 | 2875 | |||
L12146.1 | - | 3, 4 | 768 |