Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 7 | 953 | ||
NM_002718.4 | NP_002709.2 | 8 | 3388 | |||
NM_181897.2 | NP_871626.1 | 7 | 1243 | |||
XM_006713686.3 | XP_006713749.1 | 8 | 3799 | |||
XM_011512956.2 | XP_011511258.1 | 8 | 3563 | |||
XM_017006787.1 | XP_016862276.1 | 7 | 3121 | |||
GenBank mRNA | AK293012.1 | - | 8 | 3368 | ||
AK298059.1 | - | 7 | 882 | |||
AK299544.1 | - | 7 | 913 | |||
AK302163.1 | - | 7 | 785 | |||
AK314243.1 | - | 8 | 3316 | |||
AK316258.1 | - | 7 | 1196 | |||
AL389975.1 | - | 5 | 471 | |||
BC065531.1 | - | 8 | 3335 | 6161260 | ||
L07590.1 | - | 8 | 3275 | |||
L12146.1 | - | 7 | 1168 |