Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172871.2 | NP_766459.2 | 1 | 987 | ||
GenBank mRNA | AK033231.1 | - | 1 | 945 | ||
AK049803.1 | - | 1 | 942 | |||
AK129338.1 | - | 1 | 889 | |||
BC094556.1 | - | 1 | 929 | 6811241 |