Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173774.3 | NP_776135.2 | 2 | 456 | ||
GenBank mRNA | AK082651.1 | - | 448 | |||
AK134677.1 | - | 2 | 380 | |||
AK134683.1 | - | 2 | 446 | |||
AK158002.1 | - | 2 | 446 | |||
BC051188.1 | - | 2 | 416 | |||
CX241103.1 | - | 2 | 374 |