Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 3 | 829 | ||
NM_001282180.1 | NP_001269109.1 | 3 | 832 | |||
NM_001282181.1 | NP_001269110.1 | 2 | 199 | |||
NM_001282182.1 | NP_001269111.1 | 2 | 154 | |||
NM_006246.3 | NP_006237.1 | 3 | 832 | |||
GenBank mRNA | AB209811.1 | - | 3 | 785 | ||
AB451233.1 | - | 2 | 242 | |||
AB451358.1 | - | 2 | 242 | |||
AK299520.1 | - | 3 | 769 | |||
AK301796.1 | - | 2 | 199 | |||
AK313500.1 | - | 3 | 685 | |||
AK316430.1 | - | 2 | 153 | |||
BC034994.1 | - | 4 | 826 | |||
BC048305.1 | - | 3 | 565 | |||
BC064358.1 | - | 3 | 812 | |||
BC073873.1 | - | 3 | 784 | |||
BC092477.1 | - | 3 | 802 | 5267163 | ||
BC093766.1 | - | 3 | 300 | 7939611 | ||
BC101479.1 | - | 3 | 300 | 8068985 | ||
BC143231.1 | - | 3 | 297 | 9051732 | ||
BC143234.1 | - | 3 | 300 | 9051735 | ||
L76703.1 | - | 3 | 809 | |||
Z69029.1 | - | 3 | 747 |