Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 2 | 698 | ||
NM_001282180.1 | NP_001269109.1 | 2 | 701 | |||
NM_006246.3 | NP_006237.1 | 2 | 701 | |||
NR_104104.1 | 2 | 701 | ||||
GenBank mRNA | AB209811.1 | - | 2 | 654 | ||
AB451233.1 | - | 1 | 111 | |||
AB451358.1 | - | 1 | 111 | |||
AK299520.1 | - | 2 | 638 | |||
AK313500.1 | - | 2 | 554 | |||
BC013154.1 | - | 2 | 694 | |||
BC034994.1 | - | 3 | 695 | |||
BC048305.1 | - | 2 | 434 | |||
BC064358.1 | - | 2 | 681 | |||
BC073873.1 | - | 2 | 653 | |||
BC092477.1 | - | 2 | 671 | 5267163 | ||
BC093766.1 | - | 2 | 169 | 7939611 | ||
BC101479.1 | - | 2 | 169 | 8068985 | ||
BC143231.1 | - | 2 | 166 | 9051732 | ||
BC143234.1 | - | 2 | 169 | 9051735 | ||
L76703.1 | - | 2 | 678 | |||
Z69029.1 | - | 2 | 616 |