Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081203.1 | NP_001074672.1 | 21 | 2918 | ||
GenBank mRNA | AB188811.1 | - | 20 | 2847 | ||
AK133096.1 | - | 13 | 1742 | |||
AK143120.1 | - | 3 | 231 | |||
BC027253.1 | - | 3 | 306 | |||
BC138418.1 | - | 21 | 2975 | 8861438 |