Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172888.3 | NP_766476.2 | 1 | 1802 | ||
GenBank mRNA | AK035222.1 | - | 1 | 1803 | ||
AY115568.1 | - | 1 | 1759 | |||
AY283179.1 | - | 1 | 1759 | |||
BC125635.1 | - | 1 | 1673 | 40130150 |