Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134743.1 | NP_001128215.1 | 2 | 1568 | ||
NM_001282102.1 | NP_001269031.1 | 2 | 1568 | |||
NM_178731.5 | NP_848846.3 | 2 | 1322 | |||
GenBank mRNA | AK032470.1 | - | 1 | 1218 | ||
AK038656.1 | - | 2 | 1399 | |||
AY182031.1 | - | 1 | 1262 | |||
BC037216.1 | - | 2 | 1490 | 5361681 |