Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175526.3 | NP_780735.2 | 3 | 544 | ||
GenBank mRNA | AK031121.1 | - | 3 | 366 | ||
AK036846.1 | - | 3 | 399 | |||
AK049608.1 | - | 3 | 291 | |||
AK165920.1 | - | 3 | 403 | |||
BC052840.1 | - | 3 | 403 | 30050488 |