Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153590.3 | NP_705818.3 | 2 | 176 | ||
GenBank mRNA | AF306663.1 | - | 2 | 176 | ||
AY100458.1 | - | 2 | 569 | |||
BC145177.1 | - | 2 | 211 | |||
BG145575.1 | - | 2 | 154 |