Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123355.1 | NP_001116827.1 | 7 | 873 | ||
NM_001123369.1 | NP_001116841.1 | 5 | 696 | |||
NM_002721.4 | NP_002712.1 | 6 | 762 | |||
XM_011518847.2 | XP_011517149.1 | 6 | 559 | |||
GenBank mRNA | AF035158.2 | - | 6 | 535 | ||
AK295190.1 | - | 7 | 738 | |||
AK295752.1 | - | 4 | 374 | |||
AK299087.1 | - | 5 | 560 | |||
AK312332.1 | - | 6 | 594 | |||
AK316527.1 | - | 4 | 408 | |||
BC006990.1 | - | 6 | 609 | 3684339 | ||
BT019708.1 | - | 6 | 541 | |||
KU178271.1 | - | 6 | 541 | |||
KU178272.1 | - | 4 | 333 | |||
X92972.2 | - | 6 | 562 |