Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123355.1 | NP_001116827.1 | 5 | 649 | ||
NM_001123369.1 | NP_001116841.1 | 3 | 472 | |||
NM_002721.4 | NP_002712.1 | 4 | 538 | |||
XM_011518847.2 | XP_011517149.1 | 4 | 335 | |||
GenBank mRNA | AF035158.2 | - | 4 | 311 | ||
AK295190.1 | - | 5 | 514 | |||
AK299087.1 | - | 3 | 336 | |||
AK312332.1 | - | 4 | 370 | |||
BC006990.1 | - | 4 | 385 | 3684339 | ||
BT019708.1 | - | 4 | 317 | |||
DB037210.1 | - | 4 | 538 | |||
DC334216.1 | - | 5 | 512 | |||
DC369815.1 | - | 3 | 336 | |||
KU178271.1 | - | 4 | 317 | |||
X92972.2 | - | 4 | 338 |