Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172385.2 | NP_759017.1 | 6 | 3081 | ||
GenBank mRNA | AK030633.1 | - | 6 | 3082 | ||
AK034508.1 | - | 6 | 3025 | |||
AK042625.1 | - | 3 | 1141 | |||
AK122270.1 | - | 7 | 3088 | |||
BC033594.1 | - | 6 | 3003 | 5367892 |