Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglecg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172900.3 | NP_766488.2 | 3 | 530 | ||
GenBank mRNA | AK036698.1 | - | 3 | 544 | ||
AK042488.1 | - | 3 | 531 | |||
AK156766.1 | - | 3 | 572 | |||
AK162797.1 | - | 2 | 475 | |||
AY210400.1 | - | 2 | 385 | |||
BC114981.1 | - | 3 | 535 |