Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198858.1 | NP_942155.1 | 3 | 550 | ||
GenBank mRNA | AB125661.1 | - | 3 | 550 | ||
AK145438.1 | - | 622 | ||||
AY363390.1 | - | 3 | 516 | |||
BC120763.1 | - | 3 | 542 | 40129686 | ||
BC120765.1 | - | 3 | 542 | 40129688 |