Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175534.3 | NP_780743.1 | 2 | 622 | ||
GenBank mRNA | AJ320231.1 | - | 2 | 333 | ||
AK048528.1 | - | 2 | 440 | |||
AK133657.1 | - | 2 | 624 | |||
AK154886.1 | - | 2 | 417 | |||
BC096766.1 | - | 2 | 418 | 30102203 | ||
BC138305.1 | - | 2 | 331 | 8861325 | ||
BC138308.1 | - | 2 | 331 | 8861328 |
Set Membership: |
G-protein coupled receptor |