Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183026.2 | NP_898847.1 | 2 | 191 | ||
GenBank mRNA | AJ578468.2 | - | 2 | 162 | ||
BC116198.1 | - | 1 | 814 | |||
BC116199.1 | - | 2 | 162 | 40090056 |