Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005973.4 | NP_005964.3 | 6 | 1656 | ||
XM_005245313.2 | XP_005245370.1 | 7 | 1752 | |||
XM_005245314.1 | XP_005245371.1 | 5 | 1560 | |||
GenBank mRNA | AK126403.1 | - | 1 | 1507 | ||
AK289872.1 | - | 6 | 1596 | |||
BC004913.2 | - | 6 | 1572 | 3535413 | ||
BC010450.1 | - | 6 | 1584 | 4179105 | ||
BU942864.1 | - | 3 | 262 | |||
KU178273.1 | - | 6 | 1366 | |||
KU178274.1 | - | 7 | 1433 | |||
KU178275.1 | - | 5 | 1270 | |||
X97124.1 | - | 6 | 1556 |