Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172914.2 | NP_766502.1 | 4 | 496 | ||
GenBank mRNA | AK077130.1 | - | 4 | 497 | ||
BC048506.1 | - | 4 | 457 | |||
BC060957.1 | - | 4 | 494 | 6705542 |