Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175535.3 | NP_780744.2 | 3 | 429 | ||
GenBank mRNA | AK040756.1 | - | 3 | 429 | ||
AK046916.1 | - | 3 | 436 |