Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI118078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172923.3 | NP_766511.1 | 6 | 644 | ||
GenBank mRNA | AK036040.1 | - | 6 | 644 | ||
BC068128.1 | - | 7 | 800 | 6835202 |