Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243245.2 | NP_001230174.1 | 4 | 457 | ||
NM_001302926.1 | NP_001289855.1 | 4 | 486 | |||
NM_001302927.1 | NP_001289856.1 | 4 | 603 | |||
NM_002728.5 | NP_002719.3 | 4 | 490 | |||
GenBank mRNA | AK312195.1 | - | 4 | 479 | ||
AU135652.1 | - | 4 | 593 | |||
BC005929.1 | - | 4 | 483 | 4043815 | ||
CB997424.1 | - | 4 | 477 | |||
CR450311.1 | - | 3 | 416 | |||
DA841825.1 | - | 4 | 480 | |||
DQ846874.1 | - | 3 | 383 | |||
M35670.1 | - | 4 | 464 | |||
M36805.1 | - | 4 | 484 | |||
X14088.1 | - | 4 | 486 | |||
X65787.1 | - | 4 | 490 | |||
Y00809.1 | - | 4 | 484 | |||
Z26248.1 | - | 11 | 1273 |