Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172926.3 | NP_766514.2 | 13 | 1366 | ||
GenBank mRNA | AK046839.1 | - | 13 | 1308 | ||
AK049371.1 | - | 13 | 1345 | |||
AK082389.1 | - | 12 | 1260 | |||
BC080799.1 | - | 13 | 1351 | 4948538 |