Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102607.1 | NP_001096077.1 | 6 | 2450 | ||
NM_172927.3 | NP_766515.2 | 6 | 2485 | |||
GenBank mRNA | AK054356.1 | - | 6 | 2486 | ||
AK087828.1 | - | 6 | 2526 | |||
AM748260.1 | - | 4 | 2170 |