Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102607.1 | NP_001096077.1 | 6, 7 | 2620 | ||
NM_172927.3 | NP_766515.2 | 6, 7 | 2655 | |||
GenBank mRNA | AK054356.1 | - | 6, 7 | 2656 | ||
AK087828.1 | - | 6, 7 | 2696 | |||
AM748260.1 | - | 4, 5 | 2340 | |||
BC150695.1 | - | 5, 6 | 2439 | 9087606 |