Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU018091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004153.2 | NP_001004153.2 | 5 | 988 | ||
GenBank mRNA | AB201954.1 | - | 5 | 939 | ||
BC050126.1 | - | 5 | 867 | 30024412 | ||
BC080735.1 | - | 5 | 996 | 30112559 | ||
BC086454.1 | - | 4 | 1015 | 30460703 |