Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029868.2 | NP_001025039.1 | 5, 6 | 1200 | ||
NM_001290540.1 | NP_001277469.1 | 3, 4 | 855 | |||
GenBank mRNA | AK046101.1 | - | 3, 4 | 857 | ||
BC056462.1 | - | 5, 6 | 546 | 5703229 |