Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030307.2 | NP_001025478.1 | 12 | 1508 | ||
GenBank mRNA | AK136418.1 | - | 12 | 1293 | ||
AK144139.1 | - | 12 | 1296 | |||
AK166105.1 | - | 12 | 1284 | |||
AK167973.1 | - | 12 | 1259 | |||
BC068171.1 | - | 12 | 1264 | |||
BC099966.1 | - | 12 | 1348 | 6435181 |