Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172931.3 | NP_766519.1 | 8, 9 | 934 | ||
GenBank mRNA | AB510446.1 | - | 9, 10 | 1036 | ||
AB510447.1 | - | 1536 | ||||
AK046229.1 | - | 964 | ||||
BC117805.2 | - | 8, 9 | 934 | 40092308 |