Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290395.1 | NP_001277324.1 | 5 | 857 | ||
NM_177746.4 | NP_808414.2 | 4 | 487 | |||
GenBank mRNA | AY611031.1 | - | 4 | 462 | ||
AY611032.1 | - | 4 | 487 | |||
BC138085.1 | - | 4 | 455 | 8861104 | ||
BC138086.1 | - | 4 | 455 | 8861105 |