Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172932.4 | NP_766520.2 | 7 | 2443 | ||
GenBank mRNA | AK039018.1 | - | 7 | 2413 | ||
AK044438.1 | - | 2 | 802 | |||
BC069040.1 | - | 7 | 2822 | |||
BC150773.1 | - | 6 | 2264 | 9087684 |