Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005475.2 | NP_001005475.1 | 8 | 2145 | ||
NM_001114664.2 | NP_001108136.1 | 8 | 2909 | |||
GenBank mRNA | AB275281.1 | - | 8 | 2909 | ||
AK220345.1 | - | 2 | 180 | |||
AY451401.1 | - | 8 | 2902 | |||
CB236789.1 | - | 95 |