Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172935.4 | NP_766523.2 | 7 | 803 | ||
GenBank mRNA | AK077584.1 | - | 7 | 786 | ||
AK090009.1 | - | 7 | 805 | |||
AK154209.1 | - | 7 | 804 | |||
AK159860.1 | - | 7 | 800 | |||
AK168432.1 | - | 7 | 761 | |||
AK189145.1 | - | 2 | 86 | |||
AK200617.1 | - | 2 | 122 | |||
BC037005.1 | - | 7 | 839 | 5101453 |