Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198113.2 | NP_932781.1 | 11 | 1207 | ||
GenBank mRNA | AB099289.1 | - | 11 | 1205 | ||
AK038981.1 | - | 9 | 2421 | |||
AK148293.1 | - | 11 | 1200 | |||
AK149880.1 | - | 11 | 1228 | |||
AK164662.1 | - | 10 | 905 | |||
AK166342.1 | - | 11 | 1214 | |||
BC028922.1 | - | 11 | 1207 | 3987714 |