Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003917.4 | NP_001003917.2 | 8 | 873 | ||
NM_001288612.1 | NP_001275541.1 | 8 | 876 | |||
NM_001288613.1 | NP_001275542.1 | 8 | 1121 | |||
GenBank mRNA | AM085510.1 | - | 8 | 786 | ||
BC056488.1 | - | 8 | 1034 | |||
BC079884.1 | - | 8 | 821 | 6840050 | ||
BC082307.1 | - | 6 | 579 |