Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139151.4 | NP_631890.3 | 7 | 822 | ||
GenBank mRNA | AB041554.1 | - | 5 | 1401 | ||
AK028080.1 | - | 7 | 791 | |||
AK043812.1 | - | 7 | 790 | |||
AK220494.1 | - | 2095 |