Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291211.1 | NP_001278140.1 | 4, 5 | 743 | ||
NM_153594.3 | NP_705822.1 | 4, 5 | 743 | |||
GenBank mRNA | AB182640.1 | - | 4, 5 | 684 | ||
AK031335.1 | - | 4, 5 | 698 | |||
AK079927.1 | - | 1, 2 | 19 | |||
AK088800.1 | - | 4, 5 | 682 | |||
AK146080.1 | - | 4, 5 | 690 | |||
BC031360.1 | - | 3, 4 | 560 | 5359242 | ||
BC040385.1 | - | 4, 5 | 694 | 3499499 |