Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139227.4 | NP_631973.3 | 10 | 1800 | ||
GenBank mRNA | AF455111.1 | - | 9 | 1540 | ||
AI508474.1 | - | 1 | 12 | |||
AK046679.1 | - | 10 | 1801 | |||
BC035459.1 | - | 9 | 1453 | |||
BC137846.1 | - | 10 | 1721 | 8860865 |