Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139307.3 | NP_647468.2 | 2 | 1679 | ||
GenBank mRNA | AJ458938.1 | - | 2 | 1620 | ||
AK012169.1 | - | 2 | 1652 | |||
AK083684.1 | - | 2 | 1665 | |||
AK132325.1 | - | 2 | 1684 | |||
BC050274.1 | - | 2 | 1630 | 4235141 |