Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172575.3 | NP_766163.2 | 4 | 767 | ||
NM_178845.3 | NP_849173.2 | 3 | 389 | |||
GenBank mRNA | AK075945.1 | - | 3 | 380 | ||
AK077401.1 | - | 4 | 752 | |||
AK145060.1 | - | 3 | 385 | |||
AK156529.1 | - | 3 | 370 | |||
BC043453.1 | - | 3 | 380 | 5004865 |