Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139311.2 | NP_647472.2 | 10, 11 | 1781 | ||
GenBank mRNA | AK143477.1 | - | 6, 7 | 1224 | ||
AK155794.1 | - | 1, 2 | 84 | |||
AY050217.1 | - | 10, 11 | 1928 |