Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144946.4 | NP_659195.3 | 9 | 2450 | ||
GenBank mRNA | AF448840.1 | - | 9 | 1783 | ||
AK032510.1 | - | 7 | 1318 | |||
AK043710.1 | - | 9 | 1786 | |||
AK081325.1 | - | 9 | 2451 | |||
AY138990.1 | - | 9 | 1519 | |||
BC051145.1 | - | 9 | 1810 | 6590345 |