Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005247.2 | NP_001005247.2 | 7 | 1133 | ||
NM_001005248.2 | NP_001005248.2 | 7 | 1109 | |||
NM_001167864.1 | NP_001161336.1 | 7 | 1133 | |||
GenBank mRNA | AF534396.1 | - | 7 | 930 | ||
AF534397.1 | - | 7 | 906 | |||
AF534398.1 | - | 5 | 495 | |||
AF534399.1 | - | 7 | 930 | |||
AK129265.1 | - | 7 | 1094 | |||
AK141088.1 | - | 7 | 931 | |||
AK154612.1 | - | 7 | 951 | |||
AK157182.1 | - | 7 | 900 | |||
BC046405.1 | - | 2 | 112 | |||
BC046443.1 | - | 7 | 747 | |||
BC082542.1 | - | 7 | 1133 | 6405811 |