Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169153.1 | NP_001162624.1 | 2 | 236 | ||
NM_145634.3 | NP_663609.2 | 2 | 236 | |||
GenBank mRNA | AB292061.1 | - | 2 | 125 | ||
AF251703.1 | - | 2 | 190 | |||
AK089313.1 | - | 2 | 236 | |||
AK089465.1 | - | 2 | 235 | |||
AK154273.1 | - | 2 | 169 | |||
AK155367.1 | - | 2 | 234 | |||
AY214460.1 | - | 2 | 188 | |||
AY457047.1 | - | 2 | 125 | |||
BC057864.1 | - | 2 | 201 | 5318856 | ||
JX073136.1 | - | 2 | 211 |