Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159376.1 | NP_001152848.1 | 4, 5 | 411 | ||
NM_145744.2 | NP_665687.1 | 4, 5 | 234 | |||
GenBank mRNA | AF357887.1 | - | 4, 5 | 234 | ||
BC116841.1 | - | 4, 5 | 229 | 40126160 | ||
BC116843.1 | - | 4, 5 | 229 | 40126162 | ||
BC144711.1 | - | 3, 4 | 195 |