Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002736.2 | NP_002727.2 | 8 | 1061 | ||
XM_011516398.1 | XP_011514700.1 | 7 | 982 | |||
GenBank mRNA | AK128639.1 | - | 9 | 871 | ||
AK291441.1 | - | 8 | 912 | |||
BC075800.1 | - | 8 | 1100 | 30331962 | ||
M31158.1 | - | 8 | 1052 |